Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Monoclonal Antibody (IA6-2), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | IgD |
---|---|
Clone | IA6-2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H026T03B01A
![]() |
Invitrogen™
H026T03B01A |
100 Tests |
Each for $430.00
|
|
|||||
H026T02B01A
![]() |
Invitrogen™
H026T02B01A |
25 Tests |
Each for $194.00
|
|
|||||
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow CytometrySpecifications
IgD | |
5 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
P01880 | |
3495 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
IA6-2 | |
Flow Cytometry | |
NovaFluor Blue 510 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IGHD; immunoglobulin heavy constant delta | |
IgG2a κ | |
Affinity chromatography | |
IGHD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title