Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IgD Monoclonal Antibody (IA6-2), NovaFluor™ Blue 660-40S, eBioscience™, Invitrogen™

Catalog No. H026T02B07A
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H026T02B07A 25 Tests
H026T03B07A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H026T02B07A Supplier Invitrogen™ Supplier No. H026T02B07A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgD
Applications Flow Cytometry
Classification Monoclonal
Clone IA6-2
Concentration 0.6 μg/Test
Conjugate NovaFluor Blue 660-40S
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IGHD
Gene Accession No. P01880
Gene Alias Ig delta chain constant region; IGHD; IGHD1; IGHDELTA; immunoglobulin heavy constant delta
Gene Symbols IGHD
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3495
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.