Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Monoclonal Antibody (IA6-2), NovaFluor™ Yellow 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | IgD |
---|---|
Clone | IA6-2 |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H026T03Y08A
![]() |
LIFE TECHNOLOGIES
H026T03Y08A |
100 Tests |
Each for $430.00
|
|
|||||
H026T02Y08A
![]() |
LIFE TECHNOLOGIES
H026T02Y08A |
25 Tests |
Each for $194.00
|
|
|||||
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow CytometrySpecifications
IgD | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P01880 | |
3495 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
IA6-2 | |
Flow Cytometry | |
NovaFluor Yellow 755 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IGHD; immunoglobulin heavy constant delta | |
IgG2a κ | |
Affinity Chromatography | |
IGHD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title