Learn More
IgD Monoclonal Antibody (IA6-2), Super Bright™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$440.00
Specifications
Antigen | IgD |
---|---|
Clone | IA6-2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Super Bright 780 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
78-986-842
![]() |
Invitrogen™
78986842 |
100 Tests |
Each of 1 for $440.00
|
![]() |
|||||
Description
Description: The ISA-3 monoclonal antibody reacts with human ICOS (Inducible COStimulatory molecule), also known as H4, CRP-1 and AILIM. ICOS is a T cell specific activation molecule and a third member of the CD28/CTLA-4 family. Human ICOS has a relative molecular mass of 55-60 kDa, composed of 27 kDa and 29 kDa chains. Human ICOS on activated T cells has potent costimulatory activity for T cell activation and is required for humoral immune responses, in particular for memory B cell and plasma cell generation. ICOS binds to its ligand, B7h/B7RP-1 expressed on activated APCs (antigen presenting cells) and on a number of inflamed peripheral tissues. Plate-bound ISA-3 is costimulatory for T cells and induces production of IL-4, IL-5, IL-10 and other cytokines, but not IL-2. ISA-3 has the same reactivity pattern and characteristics as F44. ISA-3 was generated against the human ICOS antigen. C398.4A, anti-mouse ICOS/H4 (catalog No. 14-9949), was shown to cross-react with human ICOS but binds to an epitope different from ISA-3. C398.4A stains activated cells brighter than ISA-3; however, it also exhibits higher staining of non-activated human peripheral blood or isolated PBMC. To achieve the brightest staining of ICOS on activated human T cells, please use 13-9948 or 12-9948 rather than 11-9948. Applications Reported: This ISA-3 antibody has been reported for use in flow cytometric analysis.
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Specifications
IgD | |
Flow Cytometry | |
Super Bright 780 | |
Mouse | |
Human | |
3495 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
IA6-2 | |
Monoclonal | |
Liquid | |
RUO | |
P01880 | |
IGHD | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
IGHD |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.