Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Mouse anti-Human, R718, Clone: IA6-2 (also known as δ-IA6-2), BD Horizon™
Mouse Monoclonal Antibody
$170.00 - $378.00
Specifications
Antigen | IgD |
---|---|
Clone | IA6-2 (also known as δ-IA6-2) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow CytometrySpecifications
IgD | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
IGHD; Ig delta chain C region; Immunoglobulin heavy constant delta | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
IA6-2 (also known as δ-IA6-2) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human IgD | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title