Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Rabbit anti-Human, Clone: RM123, Abnova™

Catalog No. 89934325 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-325 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-325 Supplier Abnova Corporation Supplier No. MAB12816
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against native human IgD.

Specifications

Antigen IgD
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone RM123
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against native human IgD.
Dilution ELISA (0.01-0.1 ug/mLfor detection; 0.025-0.2 ug/well for capture) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias FLJ00382/FLJ46727/MGC29633
Gene Symbols IGHD
Host Species Rabbit
Immunogen Native purified human IgD.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3495
Test Specificity This antibody reacts to human IgD. No cross reactivity with human IgG, IgM, IgE, or IgA.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.