Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgDb Mouse, anti-Mouse, R718, Clone: 217-170, BD OptiBuild™

Catalog No. BDB752167 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752167 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752167 Supplier BD Biosciences Supplier No. 752167
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 217-170 monoclonal antibody specifically recognizes mouse IgD of Igh-C[b] haplotype (eg, C57BL/6, C57BL/10, SJL). It does not react with IgD of Igh-C[a] or related haplotypes (eg, BALB/c, AKR, CBA, C3H/He, C58, DBA/1, DBA/2, NZB, PL, RIII), or Igh-C[e] haplotype (eg, A). 217-170 antibody is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has not been shown to stimulate B-cell proliferation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgDb
Applications Flow Cytometry
Classification Monoclonal
Clone 217-170
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.