Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Antibody (4G7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500471V
Description
IgE Monoclonal specifically detects IgE in Human samples. It is validated for ELISA.Specifications
IgE | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGHE | |
Purified human IgE | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3497 | |
Human | |
Purified |
ELISA | |
4G7 | |
ELISA | |
IgE, immunoglobulin heavy constant epsilon | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
The antibody 4G7 reacts with human IgE; it recognizes an epitope different from the ones recognized by BE5 and 4H10 antibodies to IgE. | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction