Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Antibody (4H10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500472H
Description
IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Flow (Cell Surface)Specifications
IgE | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Flow Cytometry (Cell Surface) | |
4H10 | |
Western Blot, Flow Cytometry, ELISA, Flow (Cell Surface) | |
IgE, immunoglobulin heavy constant epsilon | |
Purified human IgE. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3497 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction