Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Antibody (4H10), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500472PEATT594
Description
IgE Monoclonal specifically detects IgE in Human samples. It is validated for Flow Cytometry.Specifications
IgE | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
IGHE | |
Purified human IgE. | |
0.1 mL | |
Primary | |
The antibody 4H10 reacts with human IgE; it recognizes an epitope different from the ones recognized by BE5 and 4G7 antibodies to IgE. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
4H10 | |
Flow Cytometry | |
IgE, immunoglobulin heavy constant epsilon | |
Mouse | |
Ion exchange chromatography | |
Adaptive Immunity, Immunology | |
3497.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction