Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Antibody (4H10), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500472PE
Description
IgE Monoclonal specifically detects IgE in Human samples. It is validated for Flow Cytometry.Specifications
IgE | |
Monoclonal | |
PE | |
IgE, immunoglobulin heavy constant epsilon | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
The antibody 4H10 reacts with human IgE; it recognizes an epitope different from the ones recognized by BE5 and 4G7 antibodies to IgE. | |
Store at 4C in the dark. |
Flow Cytometry | |
4H10 | |
Flow Cytometry | |
IGHE | |
Purified human IgE. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3497 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction