Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Antibody (BE5), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500470PECY55
Description
IgE Monoclonal specifically detects IgE in Human samples. It is validated for Flow Cytometry.Specifications
| IgE | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| IGHE | |
| Purified human IgE. | |
| 0.1 mL | |
| Primary | |
| The antibody BE5 reacts with human IgE; it recognizes an epitope different from the ones recognized by 4G7 and 4H10 antibodies to IgE. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| BE5 | |
| Flow Cytometry | |
| IgE, immunoglobulin heavy constant epsilon | |
| Mouse | |
| Ion exchange chromatography | |
| Adaptive Immunity, Immunology | |
| 3497.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction