Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Antibody (E411cc), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108338H
Description
IgE Monoclonal specifically detects IgE in Human samples. It is validated for Western Blot, ELISA.Specifications
IgE | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
IGHE | |
IgE from myelomatous human serum. | |
0.2 mL | |
Adaptive Immunity, Immunology | |
3497 | |
Human | |
Purified |
Western Blot, ELISA | |
E411 | |
Western Blot, ELISA | |
IgE, immunoglobulin heavy constant epsilon | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specific for human IgE, Fc-region specific. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction