Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgE Antibody (RM122), Biotin, Novus Biologicals™
SDP

Catalog No. NBP262027B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262027B 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262027B Supplier Novus Biologicals Supplier No. NBP262027B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgE Monoclonal specifically detects IgE in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen IgE
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone RM122
Concentration 1mg/mL
Conjugate Biotin
Dilution ELISA
Formulation PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide
Gene Alias IgE, immunoglobulin heavy constant epsilon
Gene Symbols IGHE
Host Species Rabbit
Immunogen Human IgE
Purification Method Protein A purified
Quantity 50 μg
Primary or Secondary Secondary
Gene ID (Entrez) 3497
Test Specificity This antibody reacts to human IgE. No cross reactivity with human IgG, IgM, IgD, or IgA.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.