Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IgE Monoclonal Antibody (Ige21), NovaFluor™ Blue 510, eBioscience™, Invitrogen™

Catalog No. H077T03B01A
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H077T03B01A 100 Tests
H077T02B01A 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H077T03B01A Supplier Invitrogen™ Supplier No. H077T03B01A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgE is one of the five classes of immunoglobulins found in mammals, characterized by its monomeric structure consisting of two heavy (epsilon) chains with four Ig-like constant domains and two light chains. Although IgE comprises only a small portion of the total immunoglobulin in serum, it plays a crucial role in type I hypersensitivity reactions, such as hay fever, asthma, hives, and anaphylactic shock. IgE is produced by terminally differentiated plasma B cells and basophils. It binds to Fc epsilon receptors on mast cells and basophils, leading to the release of histamine and other molecules that promote allergic responses. Crosslinking of IgE bound to these receptors induces degranulation of mast cells and basophils, resulting in allergic manifestations. In addition to its role in allergic responses, IgE is functionally important for immunity against parasites, highlighting its dual role in both protective and pathological immune processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgE
Applications Flow Cytometry
Classification Monoclonal
Clone Ige21
Concentration 0.2 μg/Test
Conjugate NovaFluor Blue 510
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IGHE
Gene Accession No. P01854
Gene Alias HCAK1; IgA1; IgG3; IGHA1; IGHA2; IGHG1; IGHG2; IGHG3; IGHG4; IGKC; IGKCD; IGLC; IGLC1; Immunoglobulin E; immunoglobulin heavy constant alpha 1; immunoglobulin heavy constant alpha 2 (A2m marker); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); immunoglobulin kappa constant; immunoglobulin lambda constant 1; ImmunoglobulinE; Km
Gene Symbols IGHE
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3497
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.