Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE Antibody (E411cc), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1108338G
Description
IgE Monoclonal specifically detects IgE in Human samples. It is validated for Western Blot, ELISA.Specifications
| IgE | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IGHE | |
| IgE from myelomatous human serum. | |
| 0.2 mL | |
| Adaptive Immunity, Immunology | |
| 3497 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| E411 | |
| Western Blot, ELISA | |
| IgE, immunoglobulin heavy constant epsilon | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specific for human IgE, Fc-region specific. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction