Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgE Rabbit anti-Human, Clone: RM122, Abnova™

Catalog No. 89934326 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-326 Supplier Abnova Corporation Supplier No. MAB12817
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against native human IgE.

Specifications

Antigen IgE
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone RM122
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against native human IgE.
Dilution ELISA (0.01-0.1 ug/mL for detection; 0.01-0.1 ug/well for capture) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Alias IgE
Gene Symbols IGHE
Host Species Rabbit
Immunogen Native purified human IgE.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3497
Test Specificity This antibody reacts to human IgE. No cross reactivity with human IgG, IgM, IgD, or IgA.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.