Learn More
Description
Spectrum™ Chemical Igepal CA-630 is a non-denaturing detergent. It is derived from octylphenol which serves as the hydrophobic part. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Concentration | 100% |
| Quantity | 100 mL |
| Packaging | Amber Glass Bottle |
| Grade | Reagent |
| Chemical Name or Material | Igepal CA to 630 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
