Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-I/IGF-1 Antibody (OTI4B12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271010B
Description
IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
IGF-I/IGF-1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
IGF1 | |
Recombinant protein fragment corresponding to amino acids 1-151 of human IGF-I (NP_001104753) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI4B12 | |
Western Blot, Immunohistochemistry | |
IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction | |
3479.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction