Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-I/IGF-1 Rabbit anti-Human, Clone: 23, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $524.00
Specifications
Antigen | IGF-I/IGF-1 |
---|---|
Clone | 23 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
IGF-I/IGF-1 Monoclonal specifically detects IGF-I/IGF-1 in Human samples. It is validated for ELISA.Specifications
IGF-I/IGF-1 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C | |
Recombinant Human IGF-I/IGF-1 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
23 | |
ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction | |
0.2 um filtered solution in PBS | |
3479 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title