Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-I R/IGF1R Antibody (1H7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $501.50
Specifications
Antigen | IGF-I R |
---|---|
Clone | 1H7 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:40 |
Applications | Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017782
![]() |
Novus Biologicals
NB100665370.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10066537
![]() |
Novus Biologicals
NB10066537 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
IGF-I R/IGF1R Monoclonal specifically detects IGF-I R/IGF1R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IGF-I R | |
1.0 mg/mL | |
Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer | |
P08069 | |
3480 | |
Purified human placental IGF-I receptor | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1H7 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:40 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
IGF1R | |
IgG1 | |
Protein G purified | |
NB100-66537 recognizes human CD221 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title