Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-I R/IGF1R Antibody (33255), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB391Z
Description
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ImmunocytochemistrySpecifications
IGF-I R/IGF1R | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry | |
33255 | |
Western Blot, Flow Cytometry, Immunohistochemistry, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Immunocytochemistry | |
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IGF-I R/IGF1R, Glu31-Asn932, Accession # P08069 | |
0.1 mL | |
Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3480 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction