Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-I R/IGF1R Antibody (IGF1R/4667), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314094IR
This item is not returnable.
View return policy
Description
IGF-I R/IGF1R Monoclonal antibody specifically detects IGF-I R/IGF1R in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
| IGF-I R/IGF1R | |
| Monoclonal | |
| DyLight 755 | |
| CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
| Recombinant fragment (around aa550-750) of human IGF-I R/IGF1R protein (exact sequence is proprietary) (Uniprot: P08069) | |
| 0.1 mL | |
| Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3480 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Flow Cytometry, Immunofluorescence | |
| IGF1R/4667 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction