Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-I R/IGF1R, p Tyr1165, p Tyr1166 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$268.00 - $426.00
Specifications
Antigen | IGF-I R (p Tyr1165, p Tyr1166) |
---|---|
Concentration | 1.0 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017849
![]() |
Novus Biologicals
NB100819830.025ML |
0.025 mL |
Each for $268.00
|
|
|||||
NB10081983
![]() |
Novus Biologicals
NB10081983 |
0.05 mL |
Each for $426.00
|
|
|||||
Description
IGF-I R/IGF1R Polyclonal specifically detects IGF-I R/IGF1R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
IGF-I R (p Tyr1165, p Tyr1166) | |
Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
RUO | |
Rat, Human, Mouse | |
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
IGF1R | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, Phospho Specific | |
P08069 | |
3480 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title