Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-II/IGF2 Antibody (110), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290372MFV500
Description
IGF-II/IGF2 Monoclonal antibody specifically detects IGF-II/IGF2 in Mouse samples. It is validated for ELISASpecifications
IGF-II/IGF2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
110 | |
ELISA | |
C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A | |
Rabbit immunized with purified, recombinant Mouse IGF-II/IGF2 (Uniprot#: P09535, Ala25-Glu91) | |
0.1 mL | |
Cell Cycle and Replication, Cytokine Research | |
3481 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction