Learn More
Description
Specifications
Specifications
| Antigen | IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 2G11 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/ml, Flow Cytometry 1 ug per million cells, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, CyTOF-ready |
| Gene Accession No. | P08169 |
| Gene Alias | 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR |
| Gene Symbols | IGF2R |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
