Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-II R/IGF2R Antibody (MEM-238), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500464IR
Description
IGF-II R/IGF2R Monoclonal specifically detects IGF-II R/IGF2R in Human, Primate samples. It is validated for Western Blot, Flow Cytometry.Specifications
| IGF-II R/Mannose 6 Phosphate Receptor (Cation independent) | |
| Monoclonal | |
| DyLight 755 | |
| 300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody MEM-238 recognizes an epitope between domains 2 and 5 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal ompartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry | |
| MEM-238 | |
| Western Blot, Flow Cytometry | |
| IGF2R | |
| Recombinant Vaccinia virus encoding CD222. | |
| 0.1 mL | |
| Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity | |
| 3482 | |
| Human, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction