Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-II R/IGF2R Antibody (MEM-238), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500464H
Description
IGF-II R/IGF2R Monoclonal antibody specifically detects IGF-II R/IGF2R in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOFSpecifications
IGF-II R/IGF2R | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Primate | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
MEM-238 | |
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR | |
Recombinant Vaccinia virus encoding CD222. | |
0.1 mL | |
Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity | |
3482 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction