Learn More
Invitrogen™ IGF1 Monoclonal Antibody (56408)

Description
In ELISAs, this antibody does not cross-react with recombinant mouse (rm) IGF-I, rmIGF-II, or recombinant human IGF-II. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | IGF1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 56408 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | IGF1 |
| Gene Accession No. | P05019 |
| Gene Alias | C730016P09Rik; class 1 insulin-like growth factor I preproprotein; class 2 insulin-like growth factor I preproprotein; H-IGF-1; IBP1; IGF; IGF1; Igf-1; IGF1A; IGFI; Igf-I; IGFIa; IGF-IA; IGF-IB; insulin growth factor-1; Insulin like growth factor; insulin like growth factor 1; insulin-like growth factor 1; insulin-like growth factor 1 (somatomedin C); insulin-like growth factor I; insulin-like growth factor I (somatomedin C); insulin-like growth factor IB; mechano growth factor; MGF; M-IGF-1; OTTHUMP00000195084; prepro-IGF-I; prepro-insulin-like growth factor I; R-IGF-1; somatomedin; Somatomedin C; Somatomedin-C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.