Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1R alpha Monoclonal Antibody (17-69), Invitrogen™

Catalog No. PIMA513807
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513807 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513807 Supplier Thermo Scientific Supplier No. MA513807

Mouse Monoclonal Antibody

MA5-13807 targets IGF-1 Receptor alpha in IA and IP applications and shows reactivity with Human, Porcine, and Rabbit samples. The MA5-13807 immunogen is iGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA.

IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of malignant cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF1R alpha
Applications Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 17-69
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Igf1r
Gene Accession No. P08069, Q29000
Gene Alias A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Symbols Igf1r
Host Species Mouse
Immunogen IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100144342, 3480, 397350
Target Species Human, Pig, Rabbit
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.