Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IGF1R alpha Monoclonal Antibody (1H7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA121540
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of malignant cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.
Specifications
IGF1R alpha | |
Monoclonal | |
0.3 mg/mL | |
PBS with no preservative; pH 7.4 | |
P08069 | |
Igf1r | |
Purified human placental IGF-1 receptors. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Functional Assay, Neutralization, Western Blot, Immunocytochemistry | |
1H7 | |
Unconjugated | |
Igf1r | |
A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Mouse | |
Protein G | |
RUO | |
3480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction