Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGF1R alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514677
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514677 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514677 Supplier Invitrogen™ Supplier No. PA514677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of malignant cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF1R alpha
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Igf1r
Gene Accession No. P08069
Gene Alias A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Symbols Igf1r
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 19-51 amino acids from the N-terminal region of human IGF1 Receptor (IGF1R).
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.