Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1R beta Monoclonal Antibody (1-2), Invitrogen™

Catalog No. PIMA513822
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513822 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513822 Supplier Thermo Scientific Supplier No. MA513822

Mouse Monoclonal Antibody

MA5-13822 targets IGF-1 Receptor beta in WB applications and shows reactivity with Human and Rat samples. The MA5-13822 immunogen is a 15-mer peptide corresponding to aa 1323-1337 (Tyr-RKNERALPLPQSSTC) from the C-terminal of beta-subunit of human IGF-1R.

IGF1R beta Monoclonal antibody specifically detects IGF1R beta in Human, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF1R beta
Applications Western Blot
Classification Monoclonal
Clone 1-2
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene IGF1R
Gene Accession No. P08069, P24062
Gene Alias A330103N21Rik; CD221; D930020L01; hyft; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Symbols IGF1R
Host Species Mouse
Immunogen A 15-mer peptide corresponding to aa 1323-1337 (Tyr-RKNERALPLPQSSTC) from the C-terminal of beta-subunit of human IGF-1R
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25718, 3480
Target Species Human, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.