Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF1R/Insulin R Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31668520UL
This item is not returnable.
View return policy
Description
IGF1R/Insulin R Polyclonal antibody specifically detects IGF1R/Insulin R in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
IGF1R/Insulin R | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
CD220, CD221, HHF5, IGF-I receptor, INSR, insulin receptor, insulin-like growth factor 1 receptor, IR, JTK13 | |
A synthetic phosphorylated peptide around Y1165 & Y1166 of human IGF1R/Insulin R (NP_000199.2). | |
20 μg | |
Protein Phosphatase | |
3480 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction