Learn More
Promega IGF1R Kinase Enzyme System, 1mg
Catalog No. PRV3582 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3582 1 mg
1 options
Catalog No. PRV3582 Supplier Promega Supplier No. V3582
Add to Cart
Add to Cart
Request a Quote
Recombinant human IGF1R was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. IGF1R (Insulin-like Growth Factor 1) is a transmembrane tyrosine kinase receptor that is activated by IGF-1 and by the related growth factor IGF-2.
  • Easily Screen and Profile IGF1R Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C