Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2AS Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983889
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3889 Supplier Bioss Supplier No. BS15568R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The insulin-like growth factors possess growth-promoting activity. In vitro, they are potent mitogens for cultured cells. IGF-II is influenced by placental lactogen and may play a role in fetal development. Preptin undergoes glucose-mediated co-secretion with insulin, and acts as physiological amplifier of glucose-mediated insulin secretion. Exhibits osteogenic properties by increasing osteoblast mitogenic activity through phosphoactivation of MAPK1 and MAPK3.

Specifications

Antigen IGF2AS
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IGF2-AS
Gene Accession No. Q6U949
Gene Alias IGF; IGF2 antisense RNA; IGF2AS; IGF2-AS; IGF2-AS1; Insulin like growth factor; PEG8; PEG8/IGF2AS protein
Gene Symbols IGF2-AS
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IGF2AS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51214
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.