Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGF2BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584542 Supplier Invitrogen™ Supplier No. PA584542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HALKVSYIPD EQIAQGPENG RRGGFGSRGQ PRQGSPVAAG APAKQQQVDI Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 96%.

IGF2BP1 is a member of the insulin-like growth factor 2 mRNA-binding protein family. The protein contains four K homology domains and two RNA recognition motifs. It functions by binding to the mRNAs of certain genes, including insulin-like growth factor 2, beta-actin and beta-transducin repeat-containing protein, and regulating their translation. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF2BP1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGF2BP1
Gene Accession No. Q9NZI8
Gene Alias AL024068; AW549074; B-actin zipcode-binding protein 1; coding region determinant binding protein; coding region determinant-binding protein; CRDBP; CRD-BP; D030026A21Rik; D11Moh40e; D11Moh45; fb96d05; IGF2 mRNA-binding protein 1; Igf2bp1; IGF-II mRNA-binding protein 1; Imp1; IMP-1; insulin like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 binding protein 1; insulin-like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 mRNA binding protein 1 deltaN CRDBP; insulin-like growth factor 2 mRNA-binding protein 1; insulin-like growth factor 2, binding protein 1; mir-3063; Neilsen; rZBP-1; VICKZ family member 1; Vickz1; VIon ChannelKZ1; wu:fb18f03; wu:fb96d05; wu:fi72a06; Zbp1; ZBP-1; zgc:152963; zipcode binding protein 1; zipcode-binding protein 1; zipcode-binding protein-1
Gene Symbols IGF2BP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IGF2BP1. Recombinant protein control fragment (Product #RP-103441).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10642
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.