Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGF2BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA590272

Catalog No. PIPA590272


Only null left
Add to Cart

Description

Description

Positive Samples: NIH/3T3, HeLa, A549, Jurkat, Mouse testis, Mouse kidney, Rat testis; Cellular Location: Cell projection, Cytoplasm, Nucleus, axon, dendrite, dendritic spine, filopodium, growth cone, lamellipodium, perinuclear region Immunogen sequence: NFFGPKEEVK LETHIRVPAS AAGRVIGKGG KTVNELQNLT AAEVVVPRDQ TPDENDQVIV KIIGHFYASQ MAQRKIRDIL AQVKQQHQKG QSNQAQARRK.

IGF2BP1 is a member of the insulin-like growth factor 2 mRNA-binding protein family. The protein contains four K homology domains and two RNA recognition motifs. It functions by binding to the mRNAs of certain genes, including insulin-like growth factor 2, beta-actin and beta-transducin repeat-containing protein, and regulating their translation. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IGF2BP1
Polyclonal
Unconjugated
IGF2BP1
AL024068; AW549074; B-actin zipcode-binding protein 1; coding region determinant binding protein; coding region determinant-binding protein; CRDBP; CRD-BP; D030026A21Rik; D11Moh40e; D11Moh45; fb96d05; IGF2 mRNA-binding protein 1; Igf2bp1; IGF-II mRNA-binding protein 1; Imp1; IMP-1; insulin like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 binding protein 1; insulin-like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 mRNA binding protein 1 deltaN CRDBP; insulin-like growth factor 2 mRNA-binding protein 1; insulin-like growth factor 2, binding protein 1; mir-3063; Neilsen; rZBP-1; VICKZ family member 1; Vickz1; VIon ChannelKZ1; wu:fb18f03; wu:fb96d05; wu:fi72a06; Zbp1; ZBP-1; zgc:152963; zipcode binding protein 1; zipcode-binding protein 1; zipcode-binding protein-1
Rabbit
Affinity Chromatography
RUO
10642, 140486, 303477
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88477, Q8CGX0, Q9NZI8
IGF2BP1
A synthetic peptide corresponding to a sequence within amino acids 478-577 of human IGF2BP1/IMP1 (NP_0065373).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.