Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733950 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3950 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3950 Supplier Proteintech Group Inc Supplier No. 228031AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAST3, also named as KIAA0561, belongs to the protein kinase superfamily and AGC Ser/Thr protein kinase family. MAST3 catalyze the reaction: ATP + a protein = ADP + a phosphoprotein.

Specifications

Antigen IGF2BP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IGF2BP1
Gene Accession No. O88477, Q8CGX0, Q9NZI8
Gene Alias CRD BP, CRDBP, IGF II mRNA binding protein 1, IGF2 mRNA binding protein 1, IGF2BP1, IMP 1, IMP1, VICKZ family member 1, VICKZ1, ZBP 1, ZBP1, Zip code binding protein 1, Zipcode binding protein 1
Gene Symbols IGF2BP1
Host Species Rabbit
Immunogen IGF2BP1 Fusion Protein Ag18859
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10642, 140486, 303477
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.