Learn More
Invitrogen™ IGF2BP1 Recombinant Rabbit Monoclonal Antibody (18H5L31)
Description
This antibody is predicted to react with Monkey, Dog, Cat. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | IGF2BP1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 18H5L31 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | IGF2BP1 |
| Gene Accession No. | O88477, Q9NZI8 |
| Gene Alias | AL024068; AW549074; B-actin zipcode-binding protein 1; coding region determinant binding protein; coding region determinant-binding protein; CRDBP; CRD-BP; D030026A21Rik; D11Moh40e; D11Moh45; fb96d05; IGF2 mRNA-binding protein 1; Igf2bp1; IGF-II mRNA-binding protein 1; Imp1; IMP-1; insulin like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 binding protein 1; insulin-like growth factor 2 mRNA binding protein 1; insulin-like growth factor 2 mRNA binding protein 1 deltaN CRDBP; insulin-like growth factor 2 mRNA-binding protein 1; insulin-like growth factor 2, binding protein 1; mir-3063; Neilsen; rZBP-1; VICKZ family member 1; Vickz1; VIon ChannelKZ1; wu:fb18f03; wu:fb96d05; wu:fi72a06; Zbp1; ZBP-1; zgc:152963; zipcode binding protein 1; zipcode-binding protein 1; zipcode-binding protein-1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.