Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF2BP2 Monoclonal Antibody (OTI1F9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501270S


Description
IGF2BP2 Monoclonal antibody specifically detects IGF2BP2 in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
IGF2BP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y6M1 | |
Mouse | |
Affinity chromatography | |
RUO | |
10644, 478662 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F9 | |
Unconjugated | |
IGF2BP2 | |
C330012H03Rik; Hepatocellular carcinoma autoantigen p62; IGF2 mRNA-binding protein 2; IGF2BP2; IGF-II mRNA-binding protein 2; Imp2; IMP-2; insulin like growth factor 2 mRNA binding protein 2; insulin-like growth factor 2 mRNA binding protein 2; insulin-like growth factor 2 mRNA-binding protein 2; insulin-like growth factor 2, binding protein 2; Neilsen; p62; RGD1305614; VICKZ family member 2; VICKZ2 | |
Full length human recombinant protein of human IGF2BP2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction