Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP2 Monoclonal Antibody (OTI2H7), TrueMAB™, OriGene
SDP

Catalog No. 501672416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-2416 100 μL
50-254-2416 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-2416 Supplier OriGene Supplier No. TA501271
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGF2BP2 is a member of the IGF-II mRNA-binding protein (IMP) family. It recruits target transcripts to cytoplasmic protein-RNA complexes. The protein contains several four KH domains and two RRM domains. It functions by binding to the 5′ UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. Autoantibodies against IGF2BP2 are detected in sera from some patients with hepatocellular carcinoma.

Specifications

Antigen IGF2BP2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2H7
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene IGF2BP2
Gene Accession No. Q9Y6M1
Gene Alias IMP-2, IMP2, VICKZ2
Gene Symbols IGF2BP2
Host Species Mouse
Immunogen Full length human recombit protein of human IGF2BP2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10644
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.