Learn More
IGF2BP2 Mouse anti-Human, Clone: OTI3C8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501274

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IGF2BP2 is a member of the IGF-II mRNA-binding protein (IMP) family. It recruits target transcripts to cytoplasmic protein-RNA complexes. The protein contains several four KH domains and two RRM domains. It functions by binding to the 5′ UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. Autoantibodies against IGF2BP2 are detected in sera from some patients with hepatocellular carcinoma.Specifications
IGF2BP2 | |
Monoclonal | |
Unconjugated | |
IGF2BP2 | |
IMP-2, IMP2, VICKZ2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10644 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3C8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y6M1 | |
IGF2BP2 | |
Full length human recombit protein of human IGF2BP2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.