Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGF2BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595923 Supplier Invitrogen™ Supplier No. PA595923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVEACASAEI EIMKKLREAF ENDMLAVNQQ ANLIPGLNLS ALGIFSTGLS VLSPPAGPRG APPAAPYHPF TTHSGYFSSL YPHHQFGPFP HHHSYPEQEI VNLFIPTQAV GAIIGKKGAH IKQLARFAGA SIKIAPAEGP DVSERMVIIT GPPEAQFKAQ GRIFGKLKEE NFFNPKEEVK LEAHIRVPSS TAGRVIGKGG KTVNELQNLT SAEVIVPRDQ TPDENEEVIV RIIGHFFASQ TAQRKIREIV QQVKQQEQKY PQGVASQRSK; Positive Samples: HT-29, BxPC-3, HeLa, Jurkat, NIH3T3; Cellular Location: Cytoplasm, Nucleus.

IGF2BP2 is a member of the IGF-II mRNA-binding protein (IMP) family. It recruits target transcripts to cytoplasmic protein-RNA complexes. The protein contains several four KH domains and two RRM domains. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. Autoantibodies against IGF2BP2 are detected in sera from some patients with hepatocellular carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF2BP2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IGF2BP2
Gene Accession No. Q5SF07, Q9Y6M1
Gene Alias C330012H03Rik; Hepatocellular carcinoma autoantigen p62; IGF2 mRNA-binding protein 2; IGF2BP2; IGF-II mRNA-binding protein 2; Imp2; IMP-2; insulin like growth factor 2 mRNA binding protein 2; insulin-like growth factor 2 mRNA binding protein 2; insulin-like growth factor 2 mRNA-binding protein 2; insulin-like growth factor 2, binding protein 2; Neilsen; p62; RGD1305614; VICKZ family member 2; VICKZ2
Gene Symbols IGF2BP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 330-599 of human IGF2BP2/IMP2 (NP_0065393).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10644, 303824, 319765
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.