Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IGF2R Monoclonal Antibody (MEM-238), Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIMA518132
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518132 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518132 Supplier Invitrogen™ Supplier No. MA518132
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope between amino acids 192-697 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal compartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum.

Lysosomal enzymes containing one or two mannose 6-phosphate (man6P) moieties are moved about in the cell by two distinct but interconnected cycles by means of approximately 300 kDa cation independent mannose 6-phospate receptors (CI-MPR). The biosynthetic cycle refers to the MPR transport of newly synthesized lysosomal enzymes from the trans Golgi network to late endosomes or early lysosomes. The endocytic cycle transports extracellular lysosomal enzymes from the plasma membrane via clathrin-coated vesicles to early endosomes. The entire pool of MPRs cycles between these cellular compartments every 3 hours. The steady state distribution of MPR's is predominantly within late endosomes, fewer in the trans Golgi network and approximately 10 % at cell surface. In addition to its man6P binding activity, the MPR contains a separate binding site for the type II insulin-like growth factor and is capable of binding both man6P and IGF-II simultaneously. An approximately 240 kDa soluble, truncated form, representing the extracellular domain of the protein, has also been found circulating in serum and is capable of binding both ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF2R
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-238
Conjugate Alexa Fluor 647
Formulation PBS with 0.2% BSA and 15mM sodium azide
Gene IGF2R
Gene Accession No. P11717
Gene Alias 300 kDa mannose 6-phosphate receptor; AI661837; cation-independent mannose 6-phosphate receptor; cation-independent mannose 6-phosphate receptor protein; cation-independent mannose-6-phosphate receptor; cation-independent MPR; CD222; CI Man-6-P receptor; CI-M6PR; CIMPR; CI-MPR; Igf2r; IGF-II receptor; IGF-II/CI-MPR; Igfr2; insulin like growth factor 2 receptor; insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; insulin-like growth factor receptor type 2; M6P; M6P/IGF2 receptor; M6P/IGF2R; M6PR; M6P-R; mannose-6-phosphate receptor, cation independent; mannose-6-phosphate/insulin-like growth factor II receptor; MPR 300; MPR1; MPR300; MPRI; similar to mannose-6-phosphate/insulin-like growth factor II receptor
Gene Symbols IGF2R
Host Species Mouse
Immunogen Recombinant Vaccinia virus encoding CD222.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3482
Target Species Human, Monkey
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.