Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-1 Antibody (OTI6C7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271011F
Description
IGFBP-1 Monoclonal specifically detects IGFBP-1 in Human samples. It is validated for Immunohistochemistry.Specifications
IGFBP-1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
IGFBP1 | |
Recombinant protein fragment corresponding to amino acids 26-259 of human IGFBP-1 (NP_000587) produced in E.coli. | |
0.1 mL | |
3484.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI6C7 | |
Immunohistochemistry | |
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction