Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-1 Rabbit anti-Human, Clone: S06-6J8, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | IGFBP-1 |
---|---|
Clone | S06-6J8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
IGFBP-1 Monoclonal antibody specifically detects IGFBP-1 in Human samples. It is validated for Western BlotSpecifications
IGFBP-1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Diabetes Research, Endocrinology, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
3484 | |
IgG | |
Affinity-purified |
S06-6J8 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP | |
A synthetic peptide of human IGFBP-1 (Uniprot#: P08833) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title