Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-1 Rabbit anti-Human, Mouse, Clone: 2A0B5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | IGFBP-1 |
---|---|
Clone | 2A0B5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
IGFBP-1 Monoclonal antibody specifically detects IGFBP-1 in Human, Mouse samples. It is validated for Western BlotSpecifications
IGFBP-1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Diabetes Research, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3484 | |
IgG | |
Affinity purified |
2A0B5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP | |
A synthetic peptide corresponding to a sequence within amino acids 160-259 of human IGFBP-1 (P08833). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title