Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-3 Antibody (008), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289495AF488
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
IGFBP-3 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
8 | |
ELISA, Sandwich ELISA Detection | |
acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP-3 (Gly28-Lys 291; NP_000589.2). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Prostate Cancer | |
3486 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction