Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGFBP-3 Antibody (RM0096-11B16), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122452G
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot.Specifications
IGFBP-3 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGFBP3 | |
Mouse recombinant protein of IGFBP-3 | |
0.1 mL | |
Primary | |
This antibody detects mouse IGFBP-3 in Western blotting. No cross activity to other mouse IGFBPs. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0096-11B16 | |
Western Blot | |
acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
Rat | |
Protein A or G purified | |
Apoptosis, Biologically Active Proteins, Prostate Cancer | |
3486 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction